Order Entry
United States
ContactUsLinkComponent
EcoLume™ Liquid Scintillation Cocktail, MP Biomedicals
EcoLume™ Liquid Scintillation Cocktail, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
EcoLume™ Liquid Scintillation Cocktail, MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • Storage and Handling:
    RT
  • Vapor Pressure:
    <0.1 mm Hg at 20°C
  • Description:
    EcoLume™
  • Boiling Point:
    300 °C
  • Density (at 20 °C):
    0.9 - 1.0 g/mL

Specifications

Related Information

Frequently Bought Together