Visual Scientifics Incline Plane and Car
Catalog # 470135-566
Supplier: Wards
Specifications
- Description:Visual scientifics inclined plane and car
- Cat. No.:470135-566
- Supplier no.:PTIPCR
Specifications
About this item
Visual scientifics inclined plane and car.
- Students investigate static and kinetic friction
- Twenty-Eight-Page lab manual provided
- Incline plane with track provided
- Mounting rods with hardware provided
- Car provided
In this activity, students design a simple machine to pull a car up a ramp at target speed.
28-page lab manual included, with activities and extensions, covers 15 NGSS and 17 common core standards. Platform neutral - works with your preferred probeware/video analysis tools
Certifications: Meets 15 Next Generation Science Standards concepts and 17 common core standards.
Ordering information: Visual Scientifics Base (470148-822) required, but sold separately.
Delivery: This kit includes an inclined plane track, sliding protractor and bob, car, pulley attachment, nylon cord, and mounting rods with hardware. Recommended accessories: video camera and graphical analysis software.