Visual Scientifics Backboard
470135-558
: Wards
- Description:Visual scientifics backboard
- Cat. No.:470135-558
- Supplier no.:PTBACK
This backboard mounts directly to the Visual Scientifics base, enhancing video analysis.
- Platform neutral - works with your preferred probeware/video analysis tools
- High-Contrast Grid
- Magnetic Backboard
- Mounting Hardware Provided
- Easy Installation
Students can take vertical measurements and collect data from a high-contrast grid. The backboard is magnetic and accommodates different mounting solutions for various experiments and probeware.
Mounting hardware is included. The backplace measures 2¾×16½×23½".
: Visual Scientifics Base (470148-822) required, but sold separately.