Order Entry
United States
ContactUsLinkComponent
Corning® E-Cube™ Culture System Kit, Corning
Corning® E-Cube™ Culture System Kit, Corning
Catalog # 89089-916
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
Corning® E-Cube™ Culture System Kit, Corning
Catalog # 89089-916
Supplier:  Corning
Supplier Number:  3286
Product available on GSA Advantage®
   Low Manufacturing Impact

Specifications

  • Description:
    E-Cube™ Culture System Kit (without CellCube® Module)
  • Material:
    Plastic
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    RECs, ISO 14001 - Kennebunk
  • Cat. No.:
    89089-916
  • Growth area:
    8,500 cm2
  • Supplier no.:
    3286

Specifications

About this item

The E-Cube™ System is a simple bioreactor with 8500 cm² cell growth area for growing anchorage dependent cells in only a 25.4×35.6 cm footprint. The E-Cube™ System provides a simple method to determine if cells will grow in the CellCube® Module prior to investing in the resources and funding that would be necessary for the larger, automated CellCube® System. Peristaltic pump, CO₂ incubator and medium are not provided.

Includes an oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings.

Cells grow in Corning's parallel-plate CellCube® Module on the same treated polystyrene used in Corning culture vessels. The E-Cube™ System kit consists of an oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings; CellCube® Modules must be purchased separately.