ContactUsLinkComponent
Contrex® AL, Liquid Detergent, Decon Labs
Contrex® AL, Liquid Detergent, Decon Labs
  89234-824
   Sustainable Packaging
   Waste Reducing
undefined
Contrex® AL, Liquid Detergent, Decon Labs
  89234-824
 :  5332
   Sustainable Packaging
   Waste Reducing
  • Size:
    950 mL (32.1 oz.)
  • Environmentally Preferable:
    Sustainable Packaging,
    Waste Reducing
  • Cat. No.:
    89234-824
  • Packaging:
    Poly bottle
  • Supplier no.:
    5332
Contrex® AL is a concentrated alkaline liquid detergent formulated for use in manual, soak or ultrasonic cleaning of labware, scientific apparatus and process equipment
Contrex® AL is free-rinsing, phosphate-free and disposable straight to drain. Made with recycled packaging.