Formamide, Spectral Grade
:75-12-7
:
:HCONH₂
:MFCD00007941
:13,04261
:210 °C (1013 hPa)
:2 °C
:1.1334 g/cm³ (20 °C)
:79 °C
:45.04 g/mol
:200-842-0
:GHS08
:Refrigerator
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.