ContactUsLinkComponent
Perforated Support Plate for Big Columns, Ace Glass Incorporated
Perforated Support Plate for Big Columns, Ace Glass Incorporated
Catalog # 89052-914
Supplier:  Ace Glass
Small Business Enterprise
undefined
Perforated Support Plate for Big Columns, Ace Glass Incorporated
Catalog # 89052-914
Supplier:  Ace Glass
Supplier Number:  5848-60
Small Business Enterprise
Specifications
  • Description:
    Perforated Glass Support Plate
  • Cat. No.:
    89052-914
  • Supplier no.:
    5848-60
Specifications
About this item
This 73 mm diameter perforated borosilicate glass support plate has 1×3 mm rectangular slits for supporting column packing.
Use with #80 Ace-Thred bottom adapters.