Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
BCIP/NBT
BCIP/NBT substrate (3-Components)
BCIP/NBT substrate (3-Components)
BCIP/NBT substrate (3-Components)
Catalog # 95059-176
Supplier:
KPL
CAS Number:
SDS
Expand image
BCIP/NBT substrate (3-Components)
Catalog # 95059-176
Supplier:
KPL
Supplier Number:
5420-0030
CAS Number:
SDS
Specifications
Size:
300 ml
Cat. No.:
95059-176
Supplier no.:
50-81-00
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation