About this item
LumiGLO Peroxidase Chemiluminescent substrate is an all-purpose, luminol-based substrate designed for the rapid and sensitive detection of peroxidase labeled-conjugates in Western blotting and ELISA
It enables low-picogram detection and increased sensitivity over colorimetric substrates in both applications. In blotting applications, permanent results are recorded on X-ray film. Long signal duration allows for multiple stripping and reprobing of blots. In ELISA, positive reactions are read and rapidly detected using a luminometer.
It is supplied as a 2-component, mix and use solution.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Related Information
546100.pdf
Datasheet
KPL Five Fast Fixes for Chemiluminescent Western Blot Disasters (US)