Bottom Drip Adapter for Big Columns, Ace Glass Incorporated
Catalog # 89053-028
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Bottom Drip Adapter for Big Columns
- Cat. No.:89053-028
- Supplier no.:5857-68
Specifications
About this item
This end-fitting adapter fits large columns with #80 Ace-Thred connections.
- Recessed
- Polyethylene filter disc
- Bottom outlet thread accepts 1/2-inch female NPT fitting
Adapter is recessed to allow the perforated support plate and retaining ring to fit. Bottom outlet is a thread that will accept a 1/2-inch female NPT fitting.
Ordering information: Complete item includes adapter, size -235 FETFE o-ring, 89052-914 perforated glass support plate, PTFE (89053-020) retaining ring, and 100 micron polyethylene filter disc.