Order Entry
United States
ContactUsLinkComponent
HyClone™ HEPES Buffer Solution 1 M, HyClone products (Cytiva)
HyClone™ HEPES Buffer Solution 1 M, HyClone products (Cytiva)
Catalog # 16777-032
Supplier:  Cytiva
CAS Number:  
undefined
HyClone™ HEPES Buffer Solution 1 M, HyClone products (Cytiva)
Catalog # 16777-032
Supplier:  Cytiva
Supplier Number:  SH30237.01
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    HyClone HEPES 1 M solution
  • Size:
    100 ml
  • Cat. No.:
    16777-032
  • Storage temperature:
    +2…+8 °C
  • Supplier no.:
    SH30237.01

Specifications

About this item

Maintain physiological pH in cell culture, despite changes in carbon dioxide concentration (produced by cellular respiration) with Cytiva HyClone HEPES solution.

  • 0.1 µm sterile filtered
  • pH: 7.2 - 7.5
  • Storage: 2 to 8° C
Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities