About this item
HyClone Classical Liquid Media: DMEM F12 is manufactured using ISO 9001:2000-certified processes.
- Contains 17,5 mM glucose
- 0,1 µm sterile filtered
- Manufactured using ISO 9001 certified processes
High yield cultures begin with high quality media. In order to maintain these standards, all HyClone products as well as raw material components must pass strict Quality Control testing to confirm the appropriate level of quality.
Each raw material has a specification document describing the product, the required grade or purity, the source, and the criteria for acceptance on incoming inspection. We can determine which (if any) ingredients in a medium formulation are of animal origin or, if they have come in contact with animal derived materials in manufacture.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Temperature:2...8 °C
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements