Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- CAS No.:56-75-7
- Size:500 mg
- Formula:C₁₁H₁₂Cl₂N₂O₅
- Description:2,2-Dichloro-N-[2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]acetamide
- Cat. No.:101914-578
- Supplier no.:058319-500MG
CAS #: 56-75-7
Molecular weight: 323.13
Melting point: 151°C
Molecular formula: C11H12Cl2N2O5
MDL #: MFCD00007360
Hazard code(s): Irritant