L-Alanine for synthesis
Hazard code(s): Irritant, Stored under Argon
- :56-41-7
- :C₃H₇NO₂
- :MFCD00064410
- :89.09 g/mol
- :13,00203
- :189 °C (1013 hPa)
- :314…316 °C (dec.)
- :1.4 g/cm³ (20 °C)
- :Ambient
- :200-273-8
Hazard code(s): Irritant, Stored under Argon
- :56-41-7
- :C₃H₇NO₂
- :MFCD00064410
- :89.09 g/mol
- :13,00203
- :189 °C (1013 hPa)
- :314…316 °C (dec.)
- :1.4 g/cm³ (20 °C)
- :Ambient
- :200-273-8
Product Family Options
- Size
- : 041259-25GMarketsourceMarketsource
- : 041259-100GMarketsourceMarketsource