Order Entry
United States
Orders LinkContactUsLinkComponent
3-(4-Bromophenyl)propionic acid ≥97% for synthesis

3-(4-Bromophenyl)propionic acid ≥97% for synthesis

Marketsource
Supplier:   Matrix Scientific
3-(4-Bromophenyl)propionic acid ≥97% for synthesis
Supplier:  Matrix Scientific
Marketsource
Certificates
3-(4-Bromophenyl)propionic acid ≥97% for synthesis
3-(4-Bromophenyl)propionic acid ≥97% for synthesis

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Hazard code(s): Irritant

  • CAS Number:1643-30-7
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310793
  • Molecular Weight:229.07
  • Melting Pt:133…136 °C
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 5G
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $16.10
    $16.10
    Each (5g)
       $16.10

    Supplier Number: 004941-5G
    Specifications:
    Marketsource
    Marketsource
  • 25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $55.20
    $55.20
    Each (25g)
       $55.20

    Supplier Number: 004941-25G
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.