Flask, Shaker, Three Side/Bottom Baffles, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
These long-style neck flasks have three extra-deep baffles on side and bottom to enhance gas transfer when used with rotary or reciprocating shakers
Long neck reduces splashing and is designed for polypropylene closures. Fabricated from borosilicate glass.
Ordering information: Closures not included.
Related Information
Ace Glass Cell Culture Fernbach Flasks Ace Glass Cell Culture Flasks
Product Literature
Frequently Bought Together