Flask, Nephelo Culture, Baffled, Screw Cap, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This threaded neck Nephelo culture flask has a clean-out port and a depressed sidearm
Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Fabricated from borosilicate glass.
Ordering information: Supplied with GPI #38-415 and GPI #18-415 non-toxic, rubber-lined phenolic screw caps.
Frequently Bought Together