


Ammonia solution 10% (v/v) in aqueous solution (1 + 9)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
Density:0.92 g/cm³ (20 °C)
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless liquid
- Assay (vs. Sulfuric Acid/Methyl Red):9.7 - 10.3 % (v/v)
Specification Test Results
Related Information
Frequently Bought Together