


Ammonia solution 10% (NH₃ basis)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Density:0.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear, colorless, ammonia odor
- Assay at 20 °C (traceable to NIST SRM 723):10.00 ± 0.05 % (w/v) NH3
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R0633000
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together