Silver nitrate 0.100 N (N/10) in 2-propanol
Catalog # RC7033-32
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly amber
- Size:32 oz.
- Cat. No.:RC7033-32
- Supplier no.:R7033000-1B
Specifications
Specification Test Results
- Appearance:Clear, colorless, IPA odor
- Assay at 20 °C (traceable to NIST SRM 999):0.1000 ± 0.0005 N at 20.0 °C
Specification Test Results
About this item
:
:169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:1993
ADR:3,II
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414