
Biuret reagent solution USP Test Solution (TS)
Supplier: Ricca Chemical
Certificates
About this item
:108-19-0
Storage Temperature:Ambient
UN:3266
ADR:8,II
:GHS07
Specification Test Results
- Appearance:Clear, blue, odorless
- Suitability for Use:Violet color in presence of Pepsin
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R1025000
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together