


Mercury(II) nitrate 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:10045-94-0
:
Formula:HgN₂O₆
MDL Number:MFCD00011038
:324.6
Density:1.025 g/cm³ (25 °C)
EINECS:233-152-3
UN:3287
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 999):0.1000 ± 0.0001 N at 20.0 °C
Specification Test Results
Related Information
Frequently Bought Together