Phenol red 1% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:143-74-8
:
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552
:354.38 g/mol
EINECS:205-609-7
Specification Test Results
- Appearance:clear, red, odorless
- Suitability for Use:Yellow (pH 8.2)
Specification Test Results
Frequently Bought Together