

Phenolphthalein 1% (w/v) in 70% (v/v) alcohol indicator, neutralized
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:77-09-8
:
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913
:318.33 g/mol
Boiling Pt:∼78 °C (1013 hPa)
Density:∼0.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-004-7
UN:1170
ADR:3,II
:GHS02
:GHS08
Specification Test Results
- Appearance:Clear, very light pink, alcohol odor
- Suitability for Use:Colorless (pH 8.0) - Pink (pH 10.0)
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R5618000
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together