Bromophenol blue 0.4% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:115-39-9
:
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875
:669.97 g/mol
Storage Temperature:Ambient
EINECS:204-086-2
Specification Test Results
- Appearance:Clear, dark reddish purple, odorless
- Suitability for Use:Yellow (pH 4.6)
Specification Test Results
Frequently Bought Together