ContactUsLinkComponent
Tantalum ICP Standard, 1 ml = 10 mg Ta (10000 ppm Ta)(NH₄)₂TaF₇ in H₂O 2% HF, Ricca Chemical Company
Tantalum ICP Standard, 1 ml = 10 mg Ta (10000 ppm Ta)(NH₄)₂TaF₇ in H₂O 2% HF, Ricca Chemical Company
Supplier:  Ricca Chemical
Certificates
undefined
Tantalum ICP Standard, 1 ml = 10 mg Ta (10000 ppm Ta)(NH₄)₂TaF₇ in H₂O 2% HF, Ricca Chemical Company
Supplier:  Ricca Chemical