Selenium ICP-MS Standard, 1 ml = 1 mg Se (1,000 ppm Se)Se in 3% HNO₃, Ricca Chemical Company
Catalog # RCMSSE1KN-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCMSSE1KN-100
- Element:Selenium
- Supplier no.:RMSSE1KN-100N