Scandium ICP-MS Standard, 1 ml = 1 mg Sc (1,000 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Catalog # RCMSSC1KN-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCMSSC1KN-100
- Element:Scandium
- Supplier no.:RMSSC1KN-100N