Copper ICP-MS Standard, 1 ml = 1 mg Cu (1000 ppm Cu)Cu in 3% HNO₃, Ricca Chemical Company
Catalog # RCMSCU1KN-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCMSCU1KN-100
- Element:Copper
- Supplier no.:RMSCU1KN-100N