Hafnium ICP Standard, 1 ml = 1 mg Hf (1,000 ppm Hf)HfO₂ in 3% HNO₃/ trace HF, Ricca Chemical Company
Catalog # RCPHF1KN-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCPHF1KN-100
- Element:Hafnium
- Supplier no.:RPHF1KN0-100N