Order Entry
United States
ContactUsLinkComponent
 
Boron ICP-MS Standard, 1 ml = 1 mg B (1,000 ppm B)H₃BO₃ in Water / tr NH₄OH, Ricca Chemical Company
  RCMSB1KW-100
Small Business Enterprise
 :  
undefined
Boron ICP-MS Standard, 1 ml = 1 mg B (1,000 ppm B)H₃BO₃ in Water / tr NH₄OH, Ricca Chemical Company
  RCMSB1KW-100
 :  RMSB1KW0-100N
Small Business Enterprise
 :  

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O/tr. NH₄OH
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. No.:
    RCMSB1KW-100
  • Element:
    Boron
  • Supplier no.:
    RMSB1KW0-100N