ContactUsLinkComponent
Zinc ICP Standard, 1 ml = 1 mg Zn (1,000 ppm Zn)Zn in 3% HNO₃, RICCA Chemical Company
Zinc ICP Standard, 1 ml = 1 mg Zn (1,000 ppm Zn)Zn in 3% HNO₃, RICCA Chemical Company
Supplier:  Ricca Chemical
Certificates
undefined
Zinc ICP Standard, 1 ml = 1 mg Zn (1,000 ppm Zn)Zn in 3% HNO₃, RICCA Chemical Company
Supplier:  Ricca Chemical
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.