Yttrium ICP-MS Standard, 1 ml = 10 mg Y (10000 ppm Y)Y₂O₃ in 5% HNO₃, RICCA Chemical Company
Catalog # RCMSY10KN-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:10000 µg/ml
- Matrix:5% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCMSY10KN-100
- Element:Yttrium
- Supplier no.:RMSY10KN-100N