Order Entry
United States
ContactUsLinkComponent
Terbium ICP Standard, 1 ml= 1 mg Tb (1000 ppm Tb)Tb₄O₇ in 3% HNO₃, Ricca Chemical Company
Terbium ICP Standard, 1 ml= 1 mg Tb (1000 ppm Tb)Tb₄O₇ in 3% HNO₃, Ricca Chemical Company
  RCPTB1KN-100
Small Business Enterprise
 :  
undefined
Terbium ICP Standard, 1 ml= 1 mg Tb (1000 ppm Tb)Tb₄O₇ in 3% HNO₃, Ricca Chemical Company
  RCPTB1KN-100
 :  RPTB1KN0-100N
Small Business Enterprise
 :  

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    3% HNO₃
  • Pack type:
    Poly
  • Size:
    100 ml
  • Cat. No.:
    RCPTB1KN-100
  • Element:
    Terbium
  • Supplier no.:
    RPTB1KN0-100N