Praseodymium ICP Standard, 1 mL = 1 mg Pr (1,000 ppm Pr)Pr6O11 in 3% HNO3, Ricca Chemical Company
Catalog # RCPPR1KN-100
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Pack type:Poly
- Size:100 ml
- Cat. No.:RCPPR1KN-100
- Element:Praseodymium
- Supplier no.:RPPR1KN0-100N