Order Entry
United States
ContactUsLinkComponent
 
Silver ICP-MS Standard, 1 ml = 1 mg Ag (1,000 ppm Ag)AgNO₃ in 3% HNO₃, Ricca Chemical Company
Catalog # RCMSAG1KN-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Silver ICP-MS Standard, 1 ml = 1 mg Ag (1,000 ppm Ag)AgNO₃ in 3% HNO₃, Ricca Chemical Company
Catalog # RCMSAG1KN-100
Supplier:  Ricca Chemical
Supplier Number:  RMSAG1KN-100B
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    3% HNO₃
  • Pack type:
    Poly amber
  • Size:
    100 ml
  • Cat. No.:
    RCMSAG1KN-100
  • Element:
    Silver
  • Supplier no.:
    RMSAG1KN-100B

Specifications

About this item