
Hydrochloric acid 5% (v/v) acid blank
Catalog # RCPHACID5-500
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Pack type:Poly
- Size:500 ml
- Cat. No.:RCPHACID5-500
- Supplier no.:RPHACID5-500N
Specifications
Specification Test Results
- Aluminum (Al):Actual Value Reported
- Antimony (Sb):Actual Value Reported
- Appearance:Clear, colorless, odorless
- Arsenic (As):Actual Value Reported
- Barium (Ba):Actual Value Reported
- Beryllium (Be):Actual Value Reported
- Bismuth (Bi):Actual Value Reported
- Boron (B):Actual Value Reported
- Cadmium (Cd):Actual Value Reported
- Calcium (Ca):Actual Value Reported
Specification Test Results
About this item
:
Boiling Pt:∼100 °C (1013 hPa)
Density:1.02…1.05 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,III
:36.46
EINECS:231-595-7
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324