Hydrochloric acid 0.250 N (N/4)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
:36.46
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 723):0.2500 ± 0.0001 N at 20.0 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R3625000
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together