
Potassium hydroxide 45% (w/w) in aqueous solution analytical reagent
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 723):min. 45.0% (w/w)
- Chloride (Cl):0.003% max
- Heavy Metals (as Ag):5 ppm (0.0005%) max
- Iron (Fe):2 ppm (0.0002%) max
- NH4OH Precipitate:0.005% max
- Nickel (Ni):5 ppm (0.0005%) max
- Nitrogen Compounds (as N):5 ppm (0.0005%) max
- Phosphate (PO4):2 ppm (0.0002%) max
- Potassium Carbonate K2CO3:0.2% max
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R6190000
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together