
Orthophosphoric acid 20% (v/v) in aqueous solution (1 + 4)
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7664-38-2
:
Formula:H₃PO₄
MDL Number:MFCD00011340
:98 g/mol
Density:∼1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
:GHS07
Specification Test Results
- Appearance:Clear, colorless, odorless
- Assay at 20 °C (traceable to NIST SRM 84):20.00 ± 0.04 % (v/v)
Specification Test Results
Related Information
Certificates
Search by Lot Numberghs_R5850300
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together