

Iron(III) chloride 40% (w/v) in aqueous solution
Supplier: Ricca Chemical
Certificates
About this item
CAS Number:7705-08-0
:
Formula:FeCl₃
MDL Number:MFCD00011005
:162.21 g/mol
EINECS:231-729-4
UN:2582
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear, brown, odorless
- Assay at 20 °C (traceable to NIST SRM 136):40.0 ± 0.1 % (w/v)
Specification Test Results
Related Information
Frequently Bought Together