
Hydrochloric acid 6.00 N
RC3750.1-32
Small Business Enterprise
:
- Pack type:Glass, amber
- Size:32 oz.
- Cat. No.:RC3750.1-32
- Supplier no.:R3750100-1C
- Appearance:Clear, colorless to slight yellow-green, Hydrochloric odor
- Assay at 20 °C (traceable to NIST SRM 84):6.000 ± 0.005 N at 20.0 °C
:7647-01-0
:
:HCl
:MFCD00011324
:1.06 g/cm³ (20 °C)
:Ambient
:231-595-7
:1789
:8,II
:36.46
:GHS07