ContactUsLinkComponent
BDD, Bacdown Detergent Disinfectant, Concentrated Liquid, Decon Labs, Inc.
BDD, Bacdown Detergent Disinfectant, Concentrated Liquid, Decon Labs, Inc.
Catalog # 76048-922
Supplier:  Decon Labs
undefined
BDD, Bacdown Detergent Disinfectant, Concentrated Liquid, Decon Labs, Inc.
Catalog # 76048-922
Supplier:  Decon Labs
Supplier Number:  8004
Specifications
  • Size:
    4 x 3.78 L (4 x 1 gal)
  • Cat. No.:
    76048-922
  • Packaging:
    Poly bottle
Specifications
About this item
BDD™ (Bacdown® Detergent Disinfectant) will safely and effectively clean and disinfect surfaces in labs, production areas and process equipment.  Diluted two (2) ounces per gallon of water (1:64), BDD™ is a bactericidal, virucidal, fungicidal disinfectant, effective against HIV-1, HBV and more. 
  • Concentrated quaternary (quat) disinfectant
  • Phosphate-free
Certifications: Conforms to OSHA's Bloodborne Pathogens Standard. EPA registered.