ContactUsLinkComponent
APStop™ Solution (10X Concentrate), KPL
APStop™ Solution (10X Concentrate), KPL
Catalog # 95059-226
Supplier:  KPL
CAS Number:  
APStop™ Solution (10X Concentrate), KPL
Catalog # 95059-226
Supplier:  KPL
Supplier Number:  5150-0026
CAS Number:  
Specifications
  • Description:
    APStop Solution (10X Concentrate)
  • Size:
    2 x 100 mL
  • Cat. No.:
    95059-226
  • Supplier no.:
    50-89-00
Specifications
About this item
APStop Solution (10X concentrate) is specially formulated for use with KPL colorimetric substrates for alkaline phosphatase detection in microwell applications, including BluePhos Microwell Phosphatase Substrate (Catalog Number 95059-220), FirePhos Microwell Phosphatase Substrate System (Catalog Number 95059-214) and pNPP Microwell Substrate System (Catalog number 95059-170)
  • Dilute with water and use
APStop works with all KPL AP colorimetric substrates. Plates can be read up to 2 hours after addition of APStop. Use of APStop promotes plate-to plate consistency.
APStop Solution allows the user to stop the reaction at a specific point for consistent results from assay-to-assay.