ContactUsLinkComponent
White Wooden Meter Stick
White Wooden Meter Stick
  470006-962
undefined
White Wooden Meter Stick
  470006-962
 :  W/SK CAT#
  • Color:
    White
  • Description:
    Meter stick, GLOBE approved
  • Length:
    1 m (3 ft.)
  • Material:
    Wood
  • Cat. No.:
    470006-962
  • Supplier no.:
    W/SK CAT#
A white enamel finished, high-quality hardwood meter stick, with permanently engraved scales.
One side is graduated in cm with 1 mm divisions; the other side is graduated in inches with ¹/₈ inch divisions.