ContactUsLinkComponent
White Blood Cell, Human Slide
Catalog # 470183-018
Supplier:  Avantor
undefined
White Blood Cell, Human Slide
Catalog # 470183-018
Supplier:  Avantor
Specifications
  • Description:
    White Blood Cell, Human Slide
  • Environmentally Preferable:
  • Cat. No.:
    470183-018
  • Stain:
    Giemsa
  • Supplier no.:
    WARD470183-018
  • Orientation:
    Smear
Specifications
About this item
Huge component within the immune system
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Demonstrates white blood cells and platelets. Smear (buffy coat). Giemsa.