Wax Marking Pencils
Supplier: PRECISION MOULDED POLYMERS LIMITED
Certificates
About this item
These excellent quality wax pencils are ideal for writing directly on glass, porcelain, metal, and ceramic ware. They may also be used on overhead transparencies. Reqires a sharpener, not included.
Frequently Bought Together