ContactUsLinkComponent
Ward's® Water Quality Assessment Lab Activity
Ward's® Water Quality Assessment Lab Activity
Catalog # 470030-204
Supplier:  Avantor
undefined
Ward's® Water Quality Assessment Lab Activity
Catalog # 470030-204
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Ward's® Water Quality Assessment Lab Activity
  • No. of lab groups:
    6
  • Cat. No.:
    470030-204
Specifications
About this item
So Convenient, You Don’t Even Have to Add Water
  • Kit can be reused with local water samples
  • Demonstrates the relationship between pollution and biodiversity
  • Includes a teacher's guide
Samples of clean, intermediate, and highly polluted simulated lake water are included, but you can reuse the activity for years with local water samples. By using a simple and unique analysis technique, the Palmer Algal Genera Pollution Index, your class will see that as the amount of pollution in water increases, the number of algal species decreases. The activity includes materials for six setups, a teacher’s guide, and illustrated student copymaster. Note: Includes perishable materials. Please specify delivery date.