Water Cycle Model
: Avantor
Make it Rain in your classroom!
- Includes lesson plan
Watch water evaporate from ocean surf and clouds form in the air. Students can observe numerous facets of the water cycle with this two-piece plastic terrain model, used with a lamp, water and ice to demonstrate various water-related processes. You can also use the color photos on the Study Cards to help students visualize the impact of the water cycle on our lives.
Desk Lamp required but not included
Frequently Bought Together