ContactUsLinkComponent
VWR® SoftCIDE® Extra-Mild Hand Soap
VWR® SoftCIDE® Extra-Mild Hand Soap
Catalog # 470151-086
Supplier:  Epredia
VWR® SoftCIDE® Extra-Mild Hand Soap
Catalog # 470151-086
Supplier:  Epredia
Supplier Number:  21128-04-001
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Softcide® extra-mild hand soap, refill jug (1 gal.)
  • Cat. No.:
    470151-086
  • Supplier no.:
    21128-04-601
Specifications
About this item
Unique formulation helps replenish skin’s natural oils that are rinsed away by frequent washing.
  • Replenishes natural oils
  • Extra mild on hands
  • Effective against gram-positive and gram-negative bacteria
Soap is antimicrobial strength, but extra mild on hands and contains 1% chloroxylenol (PCMX), a fast-acting broad spectrum antimicrobial ingredient that is effective against gram-negative and gram-positive bacteria, as well as yeast.